ENA341296219 Enamine

4-(5-amino-1,3,4-oxadiazol-2-yl)phenol

  • MDL number MFCD00469202
  • Cas Number : 1750-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 288-290
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92932
Availability details for
Quantity
  • Availability
  • Add to Cart