ENA341296225 Enamine

({1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene}amino)urea

  • MDL number MFCD00453968
  • Cas Number : 10281-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92939
Availability details for
Quantity
  • Availability
  • Add to Cart