ENA341296241 Enamine

3-(5-carbamoyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD06805797
  • Cas Number : 1019007-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92427
Availability details for
Quantity
  • Availability
  • Add to Cart