ENA341296248 Enamine

(6S)-6-methyl-4H,5H,6H-thieno[2,3-b]thiopyran-4-one

  • MDL number MFCD09027305
  • Cas Number : 147086-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-45
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93026
Availability details for
Quantity
  • Availability
  • Add to Cart