ENA341296255 Enamine

methyl 3-amino-2,2,3-trimethylbutanoate

  • MDL number MFCD11977705
  • Cas Number : 90886-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93049
Availability details for
Quantity
  • Availability
  • Add to Cart