ENA341296257 Enamine

1-cyanopropane-1-sulfonyl chloride

  • MDL number MFCD16080496
  • Cas Number : 27869-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93051
Availability details for
Quantity
  • Availability
  • Add to Cart