ENA341296260 Enamine

5-[(4-amino-3,5-dimethyl-1H-pyrazol-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD15976249
  • Cas Number : 1245808-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92428
Availability details for
Quantity
  • Availability
  • Add to Cart