ENA341296262 Enamine

5-(1-ethyl-1H-pyrazol-3-yl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD04970285
  • Cas Number : 1006484-70-3
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92429
Availability details for
Quantity
  • Availability
  • Add to Cart