ENA341296263 Enamine

5-[(2-amino-4-methylphenoxy)methyl]furan-2-carboxylic acid

  • MDL number MFCD15976269
  • Cas Number : 1245807-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92430
Availability details for
Quantity
  • Availability
  • Add to Cart