ENA341296264 Enamine

1-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]piperidine-3-carboxylic acid

  • MDL number MFCD06805666
  • Cas Number : 1006483-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92431
Availability details for
Quantity
  • Availability
  • Add to Cart