ENA341296265 Enamine

2-cyclopropylethan-1-amine hydrochloride

  • MDL number MFCD14525749
  • Cas Number : 89381-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-184
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93059

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-93059
pictograms GHS05,GHS07
hstatements H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart