ENA341296271 Enamine

2-(piperazin-1-yl)ethane-1-sulfonamide

  • MDL number MFCD14688720
  • Cas Number : 1247536-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93065
Availability details for
Quantity
  • Availability
  • Add to Cart