ENA341296274 Enamine

1-benzyl-3-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02231912
  • Cas Number : 360573-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92434
Availability details for
Quantity
  • Availability
  • Add to Cart