ENA341296275 Enamine

3-methoxy-4-[(phenylsulfanyl)methyl]benzaldehyde

  • MDL number MFCD02056409
  • Cas Number : 438532-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92435
Availability details for
Quantity
  • Availability
  • Add to Cart