ENA341296285 Enamine

5-chloro-3-(3-methoxyphenyl)-1,2,4-oxadiazole

  • MDL number MFCD12815769
  • Cas Number : 1179242-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93100
Availability details for
Quantity
  • Availability
  • Add to Cart