ENA341296293 Enamine

2-[(1-ethyl-1H-pyrazol-5-yl)methoxy]acetic acid

  • MDL number MFCD09859514
  • Cas Number : 1170618-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92441
Availability details for
Quantity
  • Availability
  • Add to Cart