ENA341296306 Enamine

1-(4-fluorophenyl)-3-methylbutan-2-one

  • MDL number MFCD11934894
  • Cas Number : 124258-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93124
Availability details for
Quantity
  • Availability
  • Add to Cart