ENA341296313 Enamine

tert-butyl 4-(aminomethyl)benzoate

  • MDL number MFCD04973451
  • Cas Number : 107045-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93131

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-93131
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart