ENA341296317 Enamine

1-(4-fluorophenyl)-3-methylbutan-2-ol

  • MDL number MFCD11934963
  • Cas Number : 1181600-74-7
  • Molecular Weight : 182.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-93136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93136
Availability details for
Quantity
  • Availability
  • Add to Cart