ENA341296322 Enamine

4-(chloromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09759025
  • Cas Number : 2389-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-59
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93181
Availability details for
Quantity
  • Availability
  • Add to Cart