ENA341296323 Enamine

5-[(2-aminophenoxy)methyl]furan-2-carboxylic acid

  • MDL number MFCD15976266
  • Cas Number : 1245808-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92445
Availability details for
Quantity
  • Availability
  • Add to Cart