ENA341296326 Enamine

2-amino-5-chloropyridin-4-ol

  • MDL number MFCD07787431
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-chloro-4-pyridinol
Inchi Code 1S/C5H5ClN2O/c6-3-2-8-5(7)1-4(3)9/h1-2H,(H3,7,8,9)
Inchi Key FFQWATRBGHOQBG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-93185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93185
Availability details for
Quantity
  • Availability
  • Add to Cart