ENA341296330 Enamine

N,N-dimethyl-5-[(methylamino)methyl]pyridin-2-amine

  • MDL number MFCD12403451
  • Cas Number : 179873-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93193
Availability details for
Quantity
  • Availability
  • Add to Cart