ENA341296347 Enamine

1-methyl-4-(methylcarbamoyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06806659
  • Cas Number : 1006478-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92450
Availability details for
Quantity
  • Availability
  • Add to Cart