ENA341296357 Enamine

4-(4-chloro-5-methyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD06805419
  • Cas Number : 1006495-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92453
Availability details for
Quantity
  • Availability
  • Add to Cart