ENA341296364 Enamine

2-(3-chlorophenyl)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD16320520
  • Cas Number : 1225823-53-9
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-93269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93269
Availability details for
Quantity
  • Availability
  • Add to Cart