ENA341296366 Enamine

5-[(4-aminophenoxy)methyl]furan-2-carboxylic acid

  • MDL number MFCD12856790
  • Cas Number : 1184060-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92455
Availability details for
Quantity
  • Availability
  • Add to Cart