ENA341296368 Enamine

6-(aminomethyl)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride

  • MDL number MFCD09798788
  • Cas Number : 62776-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93274
Availability details for
Quantity
  • Availability
  • Add to Cart