ENA341296369 Enamine

3-{[(5-bromofuran-2-yl)methyl]amino}-4-methylbenzoic acid

  • MDL number MFCD12040436
  • Cas Number : 1157945-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93275
Availability details for
Quantity
  • Availability
  • Add to Cart