ENA341296372 Enamine

6-bromo-N-methylpyridin-3-amine

  • MDL number MFCD00234191
  • Cas Number : 1022641-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93283
Availability details for
Quantity
  • Availability
  • Add to Cart