ENA341296374 Enamine

N'-hydroxy-2-(2-methylpropoxy)propanimidamide

  • Cas Number : 1251506-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-93285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93285
Availability details for
Quantity
  • Availability
  • Add to Cart