ENA341296375 Enamine

5-[(4-amino-3,5-dimethyl-1H-pyrazol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD15976261
  • Cas Number : 1245806-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92457
Availability details for
Quantity
  • Availability
  • Add to Cart