ENA341296380 Enamine

2-(3-phenyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD00665957
  • Cas Number : 959574-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92459
Availability details for
Quantity
  • Availability
  • Add to Cart