ENA341296384 Enamine

3-[7-(propan-2-yl)-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]propanoic acid

  • MDL number MFCD15976346
  • Cas Number : 1245808-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92461
Availability details for
Quantity
  • Availability
  • Add to Cart