ENA341296386 Enamine

2-(methoxymethyl)-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD14690989
  • Cas Number : 854754-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93304
Availability details for
Quantity
  • Availability
  • Add to Cart