ENA341296388 Enamine

2-(5-ethyl-1,3-thiazol-2-yl)ethan-1-amine

  • MDL number MFCD09904509
  • Cas Number : 1017463-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93307
Availability details for
Quantity
  • Availability
  • Add to Cart