ENA341296390 Enamine

1-(5-ethyl-1,3-thiazol-2-yl)methanamine

  • MDL number MFCD09904507
  • Cas Number : 1017435-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93309
Availability details for
Quantity
  • Availability
  • Add to Cart