ENA341296395 Enamine

3-amino-2-[(2,5-difluorophenyl)methyl]propan-1-ol

  • MDL number MFCD11639648
  • Cas Number : 1096296-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93317
Availability details for
Quantity
  • Availability
  • Add to Cart