ENA341296401 Enamine

1-{[3-(1-ethoxyethyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine

  • MDL number MFCD20502061
  • Cas Number : 1354963-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93331
Availability details for
Quantity
  • Availability
  • Add to Cart