ENA341296404 Enamine

1-methyl-3-(propan-2-yl)-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD11185460
  • Cas Number : 1094271-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93356
Availability details for
Quantity
  • Availability
  • Add to Cart