ENA341296405 Enamine

8-bromo-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD00498951
  • Cas Number : 1126824-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-93357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93357
Availability details for
Quantity
  • Availability
  • Add to Cart