ENA341296409 Enamine

2-[4-(3-chlorophenyl)piperazin-1-yl]cyclopentan-1-ol

  • MDL number MFCD13234038
  • Cas Number : 1183454-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93362
Availability details for
Quantity
  • Availability
  • Add to Cart