ENA341296411 Enamine

5-[(3-amino-1H-pyrazol-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD15976247
  • Cas Number : 1245806-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92464
Availability details for
Quantity
  • Availability
  • Add to Cart