ENA341296418 Enamine

2-[(4-amino-3-methyl-1H-pyrazol-5-yl)sulfanyl]acetic acid

  • MDL number MFCD15976390

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92466
Availability details for
Quantity
  • Availability
  • Add to Cart