ENA341296421 Enamine

1-(2-methoxyphenyl)-1H-pyrazole-4-sulfonamide

  • MDL number MFCD18057055
  • Cas Number : 1285062-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93384
Availability details for
Quantity
  • Availability
  • Add to Cart