ENA341296422 Enamine

2-phenoxyethane-1-sulfonamide

  • MDL number MFCD00754393
  • Cas Number : 100383-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93385
Availability details for
Quantity
  • Availability
  • Add to Cart