ENA341296433 Enamine

1-(difluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11558118
  • Cas Number : 1174305-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92468
Availability details for
Quantity
  • Availability
  • Add to Cart