ENA341296443 Enamine

2-amino-4H-chromene-3-carbonitrile

  • MDL number MFCD18915080
  • Cas Number : 953802-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93478
Availability details for
Quantity
  • Availability
  • Add to Cart