ENA341296444 Enamine

3-(thiophen-2-yl)piperazin-2-one

  • MDL number MFCD00867779
  • Cas Number : 86696-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93479
Availability details for
Quantity
  • Availability
  • Add to Cart