ENA341296446 Enamine

3-methylmorpholine-4-sulfonyl chloride

  • MDL number MFCD12134276
  • Cas Number : 1156821-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93482
Availability details for
Quantity
  • Availability
  • Add to Cart